Skip to Content
Merck

Skip To

03-232

Ubiquitin activating enzyme E1 (His-tag) human recombinant

Human recombinant Ubiquitin activating enzyme E1 (His-tag).

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
eCl@ss:
32160405
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

Escherichia coli, human

mol wt

Mw ~119 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

UniProt accession no.

shipped in

dry ice

General description

Human recombinant Ubiquitin activating enzyme E1 (His-tag)

Application

Human recombinant Ubiquitin activating enzyme E1 (His-tag).
Research Category
Cell Signaling

Epigenetics & Nuclear Function

Physical form

Format: Purified
In liquid containing 50 mM Tris-HCl, pH 7.5, containing 1.0 mM DTT. Frozen solution.

Preparation Note

Stable for 12 months at -80°C from date of receipt. Avoid multiple freeze/thaw cycles.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
U5633SRP6147ABE1407
manufacturer/tradename

Upstate®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

technique(s)

activity assay: suitable

technique(s)

-

technique(s)

-

technique(s)

western blot: suitable

Quality Level

100

Quality Level

200

Quality Level

-

Quality Level

100

mol wt

Mw ~119 kDa

mol wt

-

mol wt

118

mol wt

-

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice

UniProt accession no.

P22314

UniProt accession no.

P22314

UniProt accession no.

-

UniProt accession no.

P49459


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters