Skip to Content
Merck
All Photos(1)

Documents

V-902

Supelco

Vardenafil dihydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32N6O4S·2HCl
CAS Number:
Molecular Weight:
561.52
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1=C(C)N=C(CCC)N1N2)N=C2C3=CC(S(=O)(N4CCN(CC)CC4)=O)=CC=C3OCC.Cl.Cl

InChI

1S/C23H32N6O4S.2ClH/c1-5-8-20-24-16(4)21-23(30)25-22(26-29(20)21)18-15-17(9-10-19(18)33-7-3)34(31,32)28-13-11-27(6-2)12-14-28;;/h9-10,15H,5-8,11-14H2,1-4H3,(H,25,26,30);2*1H

InChI key

NOIHTGOGFDFCBN-UHFFFAOYSA-N

Gene Information

human ... PDE5A(8654)

General description

Vardenafil, a phosphodiesterase inhibitor, is used to treat erectile dysfunction. This Snap-N-Spike® reference solution is applicable for use in urine drug testing, clinical toxicology or forensic analysis by LC-MS/MS or GC/MS. Vardenafil is sold under the trade names Levitra® and Staxyn®.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Levitra is a registered trademark of Bayer Aktiengesellschaft
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Staxyn is a registered trademark of Bayer Aktiengesellschaft

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Daniel J Mans et al.
Journal of pharmaceutical and biomedical analysis, 75, 153-157 (2012-12-25)
Ion mobility spectrometry was used as a rapid screening tool for the detection of acetildenafils, sildenafils and avanafil within adulterated herbal supplement matrices. Acetildenafils show a tendency for partial fragmentation during the desorption/ionization process affording two peaks in the ion
Kenneth Beer
Journal of drugs in dermatology : JDD, 11(12), 1494-1495 (2013-02-05)
Eruptions from drugs are among the most frequently reported adverse events. The most frequent etiologic agents include antibiotics and nonsteroidal anti-inflammatory agents. These eruptions are also a frequent source of visits to dermatology of!ces. Many of the inciting agents are
[New Levitra in the treatment of erectile dysfunction].
A L Vertkin et al.
Urologiia (Moscow, Russia : 1999), (6)(6), 100-102 (2013-02-06)
Christoph Zenzmaier et al.
Endocrinology, 153(11), 5546-5555 (2012-09-06)
Phosphodiesterase type 5 (PDE5) inhibitors have been demonstrated to improve lower urinary tract symptoms secondary to benign prostatic hyperplasia (BPH). Because BPH is primarily driven by fibroblast-to-myofibroblast trans-differentiation, this study aimed to evaluate the potential of the PDE5 inhibitor vardenafil
Valter Javaroni et al.
Urology, 80(4), 858-864 (2012-08-28)
To evaluate whether the response to on-demand vardenafil could be improved by its daily usage in hypertensive men with erectile dysfunction (ED) who previously did not answer to on-demand regime. Our main efficacy criterion was per patient percentage of positive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service