Skip to Content
Merck
All Photos(1)

Key Documents

V-048

Supelco

Valeryl fentanyl hydrochloride solution

100 μg/mL in methanol (as a free base), certified reference material, ampule of 0.5 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32N2O · HCl
CAS Number:
Molecular Weight:
400.98
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 0.5 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as a free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(CCCC)N(C1=CC=CC=C1)C(CC2)CCN2CCC3=CC=CC=C3.Cl

InChI

1S/C24H32N2O.ClH/c1-2-3-14-24(27)26(22-12-8-5-9-13-22)23-16-19-25(20-17-23)18-15-21-10-6-4-7-11-21;/h4-13,23H,2-3,14-20H2,1H3;1H

InChI key

LDVFWGIGCYCMSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A certified Snap-N-Spike® solution suitable for use as starting material in calibrators and controls for valeryl fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, or forensic analysis applications. Valeryl fentanyl is a designer drug and fentanyl analog. In March, 2017, Chinese officials banned manufacturing and sale of valeryl fentanyl in China.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service