Skip to Content
Merck
All Photos(1)

Key Documents

V-019

Supelco

Cyanocobalamin (Vitamin B12) solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Vitamin B12, α-(5,6-Dimethylbenzimidazolyl)cyanocobamide, CN-Cbl, Cyanocob(III)alamin, Cyanocobalamin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C63H88CoN14O14P
CAS Number:
Molecular Weight:
1355.37
Beilstein:
4122889
EC Number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

C/C(C1=N[C@]([C@]([C@H](CC(N)=O)[C@@]/2(C)CCC(NC[C@@H](OP([O-])(O[C@H]3[C@@H](O)[C@@H](N4C=[N]5C6=C4C=C(C)C(C)=C6)O[C@@H]3CO)=O)C)=O)([H])N([Co+]5C#N)C2=C(C)/C([C@@H](CCC(N)=O)C/7(C)C)=NC7=C/8)(C)[C@@](C)(CC(N)=O)[C@@H]1CCC(N)=O)=C9N=C8[C@@H](CCC(N)=O)[C@

InChI

1S/C62H90N13O14P.CN.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-2;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);;/q;;+2/p-2/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57?,59-,60+,61+,62+;;/m1../s1

InChI key

RMRCNWBMXRMIRW-QJRSUKKJSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Cyanocobalamin is the most common and widely-produced of the vitamin B12 analogs. Vitamin B12, important for proper neuronal function and formation of blood cellular components, is monitored directly in patient serum samples by LC-MS/MS.

Application

This certified solution standard is suitable for use as a starting material in the preparation of linearity standards, calibrators or controls in a variety of testing applications from diagnostic testing of vitamin B12 deficiency by LC-MS/MS to quality control testing of vitamin B12-based dietary supplements/fortified foods or pharmaceutical preparations.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hossein Khedmat et al.
Saudi journal of kidney diseases and transplantation : an official publication of the Saudi Center for Organ Transplantation, Saudi Arabia, 24(4), 759-763 (2013-07-03)
Helicobacter pylori (HP) is a common infection worldwide and has been associated with severe morbidity. The level of vitamin B 12 in HP-infected chronic kidney disease (CKD) patients is reported to be lower than in the general population. The present
Spinal injection therapy for low back pain.
J Bart Staal et al.
JAMA, 309(23), 2439-2440 (2013-05-18)
Reversible disconnection syndrome: an unusual presentation of vitamin B12 deficiency.
Rajesh Verma et al.
Neurology India, 61(2), 167-169 (2013-05-07)
Christine M Armour et al.
Pediatrics, 132(1), e257-e261 (2013-06-19)
A neonate, who was found to have an elevated C3/C2 ratio and minimally elevated propionylcarnitine on newborn screening, was subsequently identified as having the rare cblF inborn error of vitamin B12 (cobalamin) metabolism. This disorder is characterized by the retention
Sabina Lutz et al.
Journal of the American Animal Hospital Association, 49(3), 197-203 (2013-03-29)
Juvenile cobalamin deficiency is a rare disease in border collies and its diagnosis requires a high level of clinical suspicion. The goal of this study was to increase awareness of this disease by describing the clinical and laboratory findings in

Protocols

HPLC Analysis of Vitamins B1, B6 and B12 using Ascentis® Express RP-Amide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service