Skip to Content
Merck

Skip To

O-034

Over-The-Counter Multi-Component Mixture-6 solution

100 μg/mL in acetonitrile (of each analyte), ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

(Snap-N-Spike®)

composition

Acetaminophen, 100 μg/mL , Caffeine, 100 μg/mL , Chlorpheniramine Maleate, 100 μg/mL , Ibuprofen, 100 μg/mL , Naproxen, 100 μg/mL , R,R(-)-Pseudoephedrine, 100 μg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile (of each analyte)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

multi-component solution

storage temp.

−20°C

General description

A certified Snap-N-Spike® reference solution of routinely tested over-the-counter drugs (OTCs) acetaminophen, caffeine, chlorpheniramine, ibuprofen, naproxen, and pseudoephedrine. This analytical reference standard is applicable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or pharmaceutical research applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A-053A-071D-019
description

100 μg/mL in acetonitrile (of each analyte), ampule of 1 mL, certified reference material, Cerilliant®

description

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

description

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

description

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

form

liquid

form

liquid

form

liquid

form

liquid

format

multi-component solution

format

single component solution

format

single component solution

format

single component solution

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters