Skip to Content
Merck

F-026

4-Fluorotropacocaine hydrochloride solution

1.0 mg/mL in acetonitrile: water (9:1) (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H18FNO2 · HCl
Molecular Weight:
299.77
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

1.0 mg/mL in acetonitrile: water (9:1) (as free base)

format

single component solution

storage temp.

−20°C

SMILES string

CN1[C@@]2([H])CC[C@]1([H])C[C@@H](OC(C3=CC=C(F)C=C3)=O)C2.Cl

InChI

1S/C15H18FNO2.ClH/c1-17-12-6-7-13(17)9-14(8-12)19-15(18)10-2-4-11(16)5-3-10;/h2-5,12-14H,6-9H2,1H3;1H/t12-,13+,14?;

InChI key

HDJWOWRZPURJNT-LIWIJTDLSA-N

General description

A cocaine analog first identified as a designer drug by the European Monitoring Centre for Drugs and Drug Addiction (EMCDDA) in 2008. Also called pFBT, 4-Fluorotropacocaine is often sold as an ingredient in “bath salt” products, usually mixed with other stimulant drugs such as caffeine, desoxypipradrol or synthetic cathinone derivatives. This certified solution standard is suitable for use as starting material in calibrators and controls for GC/MS or LC/MS testing methods in clinical toxicology, forensic analysis, or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F

flash_point_c

2.0 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library