Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
Quality Segment
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
1 mg/mL in methanol (as free base)
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
storage temp.
−20°C
SMILES string
O=C(C(CC)N(C)C)C1=CC(OCO2)=C2C=C1.Cl
InChI
1S/C13H17NO3.ClH/c1-4-10(14(2)3)13(15)9-5-6-11-12(7-9)17-8-16-11;/h5-7,10H,4,8H2,1-3H3;1H
InChI key
LLAFASVUPHWNTH-UHFFFAOYSA-N
General description
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description 1 mg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant® | description 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant® | description 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant® | description 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant® |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| format single component solution | format single component solution | format single component solution | format single component solution |
| application(s) forensics and toxicology | application(s) forensics and toxicology | application(s) clinical testing | application(s) clinical testing |
| manufacturer/tradename Cerilliant® | manufacturer/tradename Cerilliant® | manufacturer/tradename Cerilliant® | manufacturer/tradename Cerilliant® |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| concentration 1 mg/mL in methanol (as free base) | concentration 1.0 mg/mL in methanol (as free base) | concentration 1.0 mg/mL in methanol (as free base) | concentration 1.0 mg/mL in methanol (as free base) |
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.






