Skip to Content
Merck

D-117

cis-4,7,10,13,16,19-Docosahexaenoic acid

500 μg/mL in ethanol, certified reference material, Cerilliant®

Synonym(s):

DHA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H32O2
CAS Number:
Molecular Weight:
328.49
UNSPSC Code:
12352200
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1715505
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

500 μg/mL in ethanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCC(O)=O

InChI

1S/C22H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h3-4,6-7,9-10,12-13,15-16,18-19H,2,5,8,11,14,17,20-21H2,1H3,(H,23,24)/b4-3-,7-6-,10-9-,13-12-,16-15-,19-18-

InChI key

MBMBGCFOFBJSGT-KUBAVDMBSA-N

General description

An omega-3 fatty acid essential for normal brain growth and

function, docosahexaenoic acid (DHA) plays an important role as a signaling factor in cells for both anti- and pro-inflammatory processes. Levels of docosahexaenoic acid as well as other fatty acids are analyzed by GC/MS or LC-MS/MS methods to monitor patients undergoing diet therapy for mitochondrial or peroxisomal disorders. This Certified Spiking Solution® is suitable for use as starting material in the

preparation of linearity standards, calibrators, or controls in mass spectrometry-based DHA testing applications such as assessment of cardiovascular disease risk and fatty acid deficiency, and detection and quantification of DHA in nutraceuticals and dietary supplements.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

57.2 °F - closed cup

flash_point_c

14.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
D-117-1ML04061833557211