Skip to Content
Merck
All Photos(1)

Key Documents

D-111

Supelco

N-Desmethylzopiclone solution

100 μg/mL in acetonitrile, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15ClN6O3
CAS Number:
Molecular Weight:
374.78
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

ClC(C=N1)=CC=C1N(C2=O)C(OC(N3CCNCC3)=O)C4=C2N=CC=N4

InChI

1S/C16H15ClN6O3/c17-10-1-2-11(21-9-10)23-14(24)12-13(20-4-3-19-12)15(23)26-16(25)22-7-5-18-6-8-22/h1-4,9,15,18H,5-8H2

InChI key

CGSFZSTXVVJLIX-UHFFFAOYSA-N

General description

A certified Snap-N-Spike® solution suitable as starting material in calibrators or controls in LC/MS or GC/MS n-desmethylzopiclone testing methods for applications such as urine drug testing, clinical toxicology, or forensic analysis. N-desmethylzopiclone is a primary, inactive metabolite of zopiclone in urine and blood. The parent drug, zopiclone, is a non-benzodiazepine hypnotic used in the treatment of insomnia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service