Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C22H30O4
CAS Number:
Molecular Weight:
358.47
UNSPSC Code:
12352200
Pricing and availability is not currently available.
grade
certified reference material
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
0.5 mg/mL in acetonitrile
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
cannabis testing
format
single component solution
shipped in
wet ice
storage temp.
−20°C
General description
Cannabicyclolic Acid (CBLA) is a non-psychoactive cannabinoid found in Cannabis. This Certified Spiking Solution® is suitable for cannabicyclolic acid testing methods by GC/MS, LC/MS or HPLC for applications in testing of Cannabis potency or impurity profiling, pharmaceutical research, and forensic analysis. Cerilliant solution Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK#s for Canadian customers.
Legal Information
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
35.6 °F - closed cup
flash_point_c
2.0 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

