Skip to Content
Merck

Skip To

C-142

Cannabigerolic acid solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

CBGA solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H32O4
CAS Number:
Molecular Weight:
360.49
MDL number:
UNSPSC Code:
41116107
EC Number:
200-835-2
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

cannabis testing

format

single component solution

shipped in

dry ice

storage temp.

−70°C

SMILES string

Oc1c(c(cc(c1C(=O)O)CCCCC)O)C\C=C(\CCC=C(C)C)/C

InChI

1S/C22H32O4/c1-5-6-7-11-17-14-19(23)18(21(24)20(17)22(25)26)13-12-16(4)10-8-9-15(2)3/h9,12,14,23-24H,5-8,10-11,13H2,1-4H3,(H,25,26)/b16-12+

InChI key

SEEZIOZEUUMJME-FOWTUZBSSA-N

General description

This Certified Spiking Solution® is suitable for CBGA testing methods by HPLC, LC/MS, or GC/MS for applications in the testing of Cannabis potency or impurity profiling, pharmaceutical research, and forensic analysis. Cerilliant solution Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK#s for Canadian customers.
Cannabigerolic acid (CBGA) is one of the main cannabinoids that belongs to the class of C21 terpenophenolic compounds. A non-psychoactive cannabinoid, cannabigerolic acid (CBGA), is the precursor to THCA, CBDA, and CBCA cannabinoids in Cannabis. Cannabigerolic acid reportedly has efficacy in the treatment of cancer and schizophrenia.

Application

The Certified Reference Material (CRM) in solution can also be used as follows:
  • Multi-analysis of 11 cannabinoids in biomass and extracts of different cannabis varieties by an HPLC-UV method, following the International Conference on Harmonization (ICH) Tripartite Guideline for Validation of Analytical Procedures
  • High-pressure liquid chromatography (HPLC)- diode array detector (DAD) based quantification of eight cannabinoids in cannabis plant material
  • Determination of eight cannabinoids in hemp samples using reversed phase-high-pressure liquid chromatography (RP-HPLC) in combination with UV detector
  • Extraction of nine cannabinoids from food and beverage samples having hemp seeds, oil, and flour as ingredients followed by their analysis by liquid chromatography-electrospray ionization-QTrap-tandem mass spectrometry LC/ESI‐QTRAP‐MS/MS
  • Gas chromatography-mass spectrometry (GC-MS) based analysis of ten main cannabinoids from hemp inflorescences following their silylation and esterification for derivatization
  • Combination of UV and tandem mass spectrometric (MS/MS) detection with liquid chromatography (LC) for the analysis of major cannabinoids in 13 different cannabidiol-rich commercial products

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C-226C-153C-264
application(s)

cannabis testing

application(s)

-

application(s)

cannabis testing

application(s)

-

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

-

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

-

format

single component solution

format

single component solution

format

single component solution

format

single component solution

form

liquid

form

-

form

liquid

form

-

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

flash_point_f

55.0 °F - closed cup

flash_point_c

12.8 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters