Skip to Content
Merck
All Photos(1)

Key Documents

C-118

Supelco

Clomipramine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24Cl2N2
CAS Number:
Molecular Weight:
351.31
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CN(C)CCCN1c2ccccc2CCc3ccc(Cl)cc13

InChI

1S/C19H23ClN2.ClH/c1-21(2)12-5-13-22-18-7-4-3-6-15(18)8-9-16-10-11-17(20)14-19(16)22;/h3-4,6-7,10-11,14H,5,8-9,12-13H2,1-2H3;1H

InChI key

WIMWMKZEIBHDTH-UHFFFAOYSA-N

Gene Information

human ... SLC6A4(6532)

General description

Clomipramine is a tricyclic antidepressant used to many conditions from major depression and panic disorder to narcolepsy and obsessive compulsion disorder (OCD). This certified solution standard is suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Lagus et al.
Journal of neurogenetics, 26(3-4), 397-412 (2012-07-13)
Disturbances in sleep are encountered in the majority of patients with depressive disorder. To elucidate the molecular mechanisms behind this relationship, we examined gene expression changes in a rodent model for disturbed sleep and depression. The animals were treated with
Delphine Oudiette et al.
Consciousness and cognition, 21(3), 1129-1140 (2012-06-01)
To test whether mental activities collected from non-REM sleep are influenced by REM sleep, we suppressed REM sleep using clomipramine 50mg (an antidepressant) or placebo in the evening, in a double blind cross-over design, in 11 healthy young men. Subjects
P M Pitychoutis et al.
Neuroscience, 210, 152-167 (2012-03-24)
It is well established that women experience major depression at roughly twice the rate of men. Interestingly, accumulating clinical and experimental evidence shows that the responsiveness of males and females to antidepressant pharmacotherapy, and particularly to tricyclic antidepressants (TCAs), is
Saied Saeed Hosseiny Davarani et al.
Analytica chimica acta, 725, 51-56 (2012-04-17)
Recent advances in electromembrane extraction (EME) methodology calls for effective and accessible detection methods. Using imipramine and clomipramine as model therapeutics, this proof-of-principle work combines EME with gas chromatography analysis employing a flame ionization detector (FID). The drugs were extracted
S A Savelyev et al.
Neuroscience, 212, 149-158 (2012-04-21)
Neonatal treatment of rat pups with clomipramine (CLI) has been shown to cause long-lasting and persistent depression-related behaviors and changes in sleep architecture and in brain-derived neurotrophic factor (BDNF) signaling in adult animals, producing an animal model of depression. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service