Skip to Content

C-110

Catecholamine Mix 2 (Metanephrines) solution

1.0 mg/mL each component in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Metanephrines solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H13NO3·HCl
Molecular Weight:
219.67
UNSPSC Code:
41116107
EC Number:
200-659-6
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

(±)-Metanephrine HCl , (±)-Normetanephrine HCl

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL each component in methanol (as free base)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing
forensics and toxicology

format

multi-component solution

storage temp.

−70°C

General description

A Certified Spiking Solution® of (±)-Metanephrine HCl and (±)-Normetanephrine HCl. This multi-component mix is suitable for critical quantitative applications in research and clinical environments including: for use in preparation of controls, calibrators, and linearity standards for LC-MS/MS assays; as system tuning/suitability standards; linearity verification; and internal proficiency testing. The stable, quantitative solution format provides significant benefits to the user such as convenience of use, material savings, and increased efficiency of labor.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Analyte

Description

  • DL-Metanephrine hydrochloride

  • DL-Normetanephrine hydrochloride

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup

target_organs

Eyes,Central nervous system

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library