Skip to Content
Merck
All Photos(1)

Key Documents

C-060

Supelco

Cyclobenzaprine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22ClN
CAS Number:
Molecular Weight:
311.85
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CN(C)CC\C=C1\c2ccccc2C=Cc3ccccc13

InChI

1S/C20H21N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-14H,7,15H2,1-2H3;1H

InChI key

VXEAYBOGHINOKW-UHFFFAOYSA-N

Gene Information

General description

Cyclobenzaprine is a muscle relaxant sold under the trade name Flexeril® to relieve skeletal muscle spasms and associated pain from acute musculoskeletal conditions. This Snap-N-Spike® Reference Solution is suitable for use in LC/MS or GC/MS methods for clinical toxicology, forensic analysis, or urine drug testing applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Flexeril is a registered trademark of ALZA Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Is there sufficient evidence to suggest cyclobenzaprine might be implicated in causing serotonin toxicity?
P Ken Gillman
The American journal of emergency medicine, 27(4), 509-510 (2009-06-27)
Harvey Moldofsky et al.
The Journal of rheumatology, 38(12), 2653-2663 (2011-09-03)
To determine the effects of bedtime very low dose (VLD) cyclobenzaprine (CBP) on symptoms and sleep physiology of patients with fibromyalgia (FM), unrefreshing sleep, and the α-nonREM sleep electroencephalographic (EEG) anomaly at screening. Of 37 patients with FM in the
Lindsay T Morgan Bicknell et al.
International journal of psychiatry in medicine, 44(4), 305-314 (2012-01-01)
Lamotrigine is FDA-approved as a maintenance treatment of bipolar disorder, but its common off-label uses include bipolar depression and antidepressant augmentation in patients with major depressive disorder. Among other adverse effects, cutaneous reactions, particularly erythema multiforme, are cited as concerns
Yi Xiang et al.
Biomedical chromatography : BMC, 26(9), 1083-1088 (2011-11-29)
A sensitive, rapid and simple liquid chromatography-electrospray ionization mass spectrometry (LC-ESI-MS/MS) method was developed for the quantitative determination of cyclobenzaprine in human plasma, to study the pharmacokinetic behavior of cyclobenzaprine capsule in healthy Chinese volunteers. With escitalopram as the internal
Incidence of tricyclic antidepressant-like complications after cyclobenzaprine overdose.
Vikhyat S Bebarta et al.
The American journal of emergency medicine, 30(2), 374-374 (2011-11-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service