Skip to Content
Merck

Skip To

C-007

Codeine-D3 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H18D3NO3
CAS Number:
Molecular Weight:
302.38
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-659-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])N1CC[C@]23[C@H]4Oc5c(OC)ccc(C[C@@H]1[C@@H]2C=C[C@@H]4O)c35

InChI

1S/C18H21NO3/c1-19-8-7-18-11-4-5-13(20)17(18)22-16-14(21-2)6-3-10(15(16)18)9-12(11)19/h3-6,11-13,17,20H,7-9H2,1-2H3/t11-,12+,13-,17-,18-/m0/s1/i1D3

InChI key

OROGSEYTTFOCAN-SBGSAQJDSA-N

General description

Codeine-D3 is a stable-labeled internal standard suitable for many LC/MS or GC/MS testing applications including pain prescription monitoring, forensic analysis, clinical toxicology, urine drug testing or isotope dilution methods. Codeine is an opiate used for its analgesic, antitussive, and sedative properties and is the second-most predominant alkaloid in opium. Since opiate drugs are classified as controlled substances, Cerilliant offers most opiate reference standards as DEA-exempt solutions for added convenience.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C-040C-005C-041
application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

forensics and toxicology

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

form

liquid

form

liquid

form

liquid

form

liquid

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?

Explore all of our products under Codeine-D3 solution


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters