Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C13H9D9ClNO · HCl
CAS Number:
Molecular Weight:
285.26
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-659-6
MDL number:
Pricing and availability is not currently available.
grade
certified reference material
Quality Segment
form
liquid
feature
(Snap-N-Spike®)
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
100 μg/mL in methanol
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
clinical testing
format
single component solution
storage temp.
−20°C
SMILES string
CC(NC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C(C1=CC=CC(Cl)=C1)=O.Cl
InChI
1S/C13H18ClNO.ClH/c1-9(15-13(2,3)4)12(16)10-6-5-7-11(14)8-10;/h5-9,15H,1-4H3;1H/i2D3,3D3,4D3;
InChI key
HEYVINCGKDONRU-WWMMTMLWSA-N
General description
Bupropion is an antidepressant and smoking cessation drug sold under the trade names Wellbutrin® and Zyban®. Suitable uses for this certified solution standard include as an internal standard for quantitation of bupropion levels in biological matrices such as urine or isotope dilution methods by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis applications.
Legal Information
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Wellbutrin is a registered trademark of GlaxoSmithKline LLC
Zyban is a registered trademark of Everris International B.V.
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes
Storage Class
3 - Flammable liquids
WGK
WGK 1
Flash Point (°F)
49.5 °F
Flash Point (°C)
9.7 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


