Skip to Content

B-043

Brompheniramine maleate solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H23BrN2O4
CAS Number:
Molecular Weight:
435.31
UNSPSC Code:
41116107
MDL number:
NACRES:
NA.24
EC Number:
200-659-6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

OC(=O)\C=C/C(O)=O.CN(C)CCC(c1ccc(Br)cc1)c2ccccn2

InChI

1S/C16H19BrN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

SRGKFVAASLQVBO-BTJKTKAUSA-N

Gene Information

human ... HRH1(3269)

General description

Brompheniramine is an over-the-counter antihistamine sold under the trade names Bromfed®, Dimetapp®, and Lodrane®. The drug is indicated for treatment of the common cold and allergic rhinitis symptoms including runny nose and itchy eyes. This Snap-N-Spike® Reference Solution is suitable for use as starting material in calibrators or controls for LC/MS or GC/MS methods in clinical toxicology, forensic analysis, and pharmaceutical research.

Legal Information

Lodrane is a registered trademark of ECR Pharmaceuticals Co., Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Dimetapp is a registered trademark of Wyeth LLC
Bromfed is a registered trademark of Wockhardt EU Operations Swiss AG


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class

3 - Flammable liquids

WGK

WGK 1

Flash Point (°F)

49.5 °F - closed cup

Flash Point (°C)

9.7 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library