Skip to Content
Merck

B-037

Butorphanol tartrate solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H35NO8
Molecular Weight:
477.55
UNSPSC Code:
41116107
MDL number:
NACRES:
NA.24
EC Number:
200-659-6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O[C@@H]([C@H](O)C(O)=O)C(O)=O.Oc1ccc2C[C@H]3N(CC[C@@]4(CCCC[C@@]34O)c2c1)CC5CCC5

InChI

1S/C21H29NO2.C4H6O6/c23-17-7-6-16-12-19-21(24)9-2-1-8-20(21,18(16)13-17)10-11-22(19)14-15-4-3-5-15;5-1(3(7)8)2(6)4(9)10/h6-7,13,15,19,23-24H,1-5,8-12,14H2;1-2,5-6H,(H,7,8)(H,9,10)/t19-,20+,21-;1-,2-/m10/s1

InChI key

GMTYREVWZXJPLF-AFHUBHILSA-N

General description

Butorphanol is a synthetic opioid used as a treatment for migraines or moderate to severe pain. This Certified Snap-N-Spike® Solution is suitable for use as calibrators or controls in a variety of LC/MS or GC/MS applications from forensic analysis and urine drug testing to pain prescription monitoring and clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library