Skip to Content
Merck

B-013

Benzoylecgonine-D8 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H11NO4D8
CAS Number:
Molecular Weight:
297.38
MDL number:
UNSPSC Code:
41116107
EC Number:
200-659-6
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

InChI

1S/C16H19NO4/c1-17-11-7-8-12(17)14(15(18)19)13(9-11)21-16(20)10-5-3-2-4-6-10/h2-6,11-14H,7-9H2,1H3,(H,18,19)/t11-,12-,13+,14+/m1/s1/i1D3,2D,3D,4D,5D,6D

InChI key

GVGYEFKIHJTNQZ-GDERWKOJSA-N

General description

A stable-labeled internal standard suitable for isotope dilution methods or benzoylecgonine testing in urine, saliva, blood, or hair by GC/MS or LC/MS for applications in clinical toxicology, urine drug testing, pain prescription monitoring or forensic analysis. Benzoylecgonine is the primary metabolite of cocaine and a major biomarker for determination of cocaine use.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Benzoylecgonine-D8 solution


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library