Skip to Content
Merck

A-202

Acrolein-2,4-DNPH

certified reference material, vial of 50 mg, Cerilliant®

Synonym(s):

Acrolein-2,4-dinitrophenylhydrazone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H8N4O4
CAS Number:
Molecular Weight:
236.18
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

vial of 50 mg

manufacturer/tradename

Cerilliant®

format

neat

storage temp.

−20°C

SMILES string

[N+](=O)([O-])c1c(ccc(c1)[N+](=O)[O-])N\N=C\C=C

InChI

1S/C9H8N4O4/c1-2-5-10-11-8-4-3-7(12(14)15)6-9(8)13(16)17/h2-6,11H,1H2/b10-5+

InChI key

JJPZHGIYUVFTGG-BJMVGYQFSA-N

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Acrolein-2,4-DNPH


wgk

WGK 3

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library