Skip to Content
Merck

A-176

Glutaryl-L-Carnitine lithium salt solution

100 μg/mL (90:10 Methanol: 1% aqueous formic acid), certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H20NO6 · Li
CAS Number:
Molecular Weight:
281.23
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (90:10 Methanol: 1% aqueous formic acid)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

InChI

1S/C12H21NO6/c1-13(2,3)8-9(7-11(16)17)19-12(18)6-4-5-10(14)15/h9H,4-8H2,1-3H3,(H-,14,15,16,17)/t9-/m1/s1

InChI key

NXJAXUYOQLTISD-SECBINFHSA-N

General description

A Certified Spiking Solution® of routinely tested Glutaryl-L-Carnitine at a concentration of 100 μg/mL. Acylcarnitines are biomarker for many inborn errors of metabolism that cause several metabolic diseases and are routinely tested in newborn screening programs. This single-component solution certified reference material is suitable for critical quantitative applications in research and clinical environments including: for use in preparation of controls, calibrators, and linearity standards for LC-MS/MS assays; as system tuning/suitability standards; linearity verification; and internal proficiency testing. The stable, quantitative solution format provides significant benefits to the user such as convenience of use, material savings, and increased efficiency of labor. Certified concentration value provides increased efficiency from less time remaking pools due to dilution errors or rerunning samples.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library