Skip to Content
Merck

A-147

Acylcarnitines IS Mix 2 (C4,iC5,C6,C8,C10,C12,C14,C18) solution

100-200 μg/mL (90:10 Methanol: 1% aqueous formic acid), certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Acylcarnitine Internal Standard Mix

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19C9H28NO4
Molecular Weight:
442.53
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

Butyryl-L-Carnitine-(N-Methyl-D3), 100 μg/mL , Decanoyl-L-Carnitine-(N-Methyl-D3), 100 μg/mL , Hexanoyl-L-Carnitine-(N-Methyl-D3), 100 μg/mL , Isovaleryl-DL-Carnitine-(N,N,N-trimethyl-D9), 100 μg/mL , Lauroyl-L-Carnitine-(N,N,N-Trimethyl-D9), 100 μg/mL , Myristoyl-L-Carnitine-(N,N,N-Trimethyl-D9), 100 μg/mL , Octanoyl-L-Carnitine-(N-Methyl-D3), 100 μg/mL , Stearoyl-L-Carnitine-(N-Methyl-D3), 200 μg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100-200 μg/mL (90:10 Methanol: 1% aqueous formic acid)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

multi-component solution

storage temp.

−70°C

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library