Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
grade
certified reference material
form
liquid
feature
(Snap-N-Spike®)
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
1.0 mg/mL in methanol (as free base)
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
clinical testing
format
single component solution
storage temp.
−20°C
SMILES string
Cl.N(CCC=C1c2c(cccc2)CCc3c1cccc3)(C([2H])([2H])[2H])C
InChI
1S/C20H23N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-12H,7,13-15H2,1-2H3;1H/i1D3;
InChI key
KFYRPLNVJVHZGT-NIIDSAIPSA-N
General description
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| format single component solution | format single component solution | format single component solution | format single component solution |
| concentration 1.0 mg/mL in methanol (as free base) | concentration 100 μg/mL in methanol (as free base) | concentration 1.0 mg/mL in methanol | concentration 1.0 mg/mL in methanol (as free base) |
| form liquid | form liquid | form liquid | form liquid |
| manufacturer/tradename Cerilliant® | manufacturer/tradename Cerilliant® | manufacturer/tradename Cerilliant® | manufacturer/tradename Cerilliant® |
| feature (Snap-N-Spike®) | feature SNAP-N-SPIKE®, SNAP-N-SHOOT® | feature SNAP-N-SPIKE®, SNAP-N-SHOOT® | feature (Snap-N-Spike®) |
Still not finding the right product?
Explore all of our products under Amitriptyline-d3 hydrochloride solution
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





