Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
Quality Segment
form
liquid
feature
(Snap-N-Spike®)
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
drug control
USDEA Schedule I; Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)
concentration
100 μg/mL in methanol
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
storage temp.
−20°C
SMILES string
N2(CCC(CC2)N(c3ccccc3)C(=O)C)CCc1ccccc1
InChI
1S/C21H26N2O/c1-18(24)23(20-10-6-3-7-11-20)21-13-16-22(17-14-21)15-12-19-8-4-2-5-9-19/h2-11,21H,12-17H2,1H3
InChI key
FYIUUQUPOKIKNI-UHFFFAOYSA-N
General description
Legal Information
Disclaimer
Still not finding the right product?
Explore all of our products under Acetyl fentanyl-13C6 solution
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
49.5 °F
flash_point_c
9.7 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


