Skip to Content
Merck
All Photos(1)

Documents

880410P

Avanti

14:0 PEG350 PE

Avanti Research - A Croda Brand 880410P, powder

Synonym(s):

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-350] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (880410P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880410P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O.[NH4+]

General description

14:0 PEG350 PE is a polyethylene glycol (PEG) conjugated phospholipid. PEGylated phospholipids are amphiphilic polymers.

Application

14:0 PEG350 PE might be suitable for the preparation of floating bilayer lipid membrane.

Biochem/physiol Actions

14:0 PEG350 PE is useful in liposome formulation and used as drug carriers.

Packaging

5 mL Clear Glass Sealed Ampule (880410P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MICRO AND NANO SENSING: AN INTERNATIONAL JOURNAL, 110-110 (2014)
Cell culture studies of a carborane cholesteryl ester with conventional and PEG liposomes
Peacock GF, et al.
Drug delivery, 10(1), 29-34 (2003)
Oblique Self-Assemblies and Order- Order Transitions in Polypeptide Complexes with PEGylated Triple-Tail Lipids
Hanski S, et al.
Biomacromolecules, 11(12), 3440-3447 (2010)
Electrochemical and PM-IRRAS studies of floating lipid bilayers assembled at the Au (111) electrode pre-modified with a hydrophilic monolayer
Su Z, et al.
Journal of Electroanalytical Chemistry, 688, 76-85 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service