Skip to Content
Merck
All Photos(1)

Key Documents

870273C

Avanti

18:1 Biotinyl Cap PE

Avanti Research - A Croda Brand 870273C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(cap biotinyl) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H102N4O11PNaS
CAS Number:
Molecular Weight:
1105.47
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

liquid

packaging

pkg of 1 × 2.5 mL (870273C-25mg)
pkg of 2 × 4 mL (870273C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870273C

concentration

10 mg/mL (870273C-25mg)
25 mg/mL (870273C-200mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(CCCCCNC(CCCC[C@](SC[C@@]1(NC(N2)=O)[H])([C@]12[H])[H])=O)=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C57H103N4O11PS.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-33-41-54(64)69-46-49(72-55(65)42-34-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)47-71-73(67,68)70-45-44-59-53(63)39-32-31-37-43-58-52(62)40-36-35-38-51-56-50(48-74-51)60-57(66)61-56;/h17-20,49-51,56H,3-16,21-48H2,1-2H3,(H,58,62)(H,59,63)(H,67,68)(H2,60,61,66);/q;+1/p-1/b19-17-,20-18-;/t49-,50+,51+,56+;/m1./s1

InChI key

VQSGNVZURNOPBT-XEAQQBNDSA-M

General description

18:1 Biotinyl Cap PE, also known as 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(cap biotinyl) or DOPE-B-Cap, belongs to a class of head group modified functionalized lipids.

Application

18:1 Biotinyl Cap PE (1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(cap biotinyl) has been used in preparation of liposomes[1] and large unilamellar vesicles (LUV).[2]

Packaging

5 mL Clear Glass Sealed Ampule (870273C-200mg)
5 mL Clear Glass Sealed Ampule (870273C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideyo Takatsuki et al.
Nanotechnology, 22(24), 245101-245101 (2011-04-22)
The potential of using actin bundles for the transport of liposomes and single cells across myosin-coated surfaces is investigated. Compared to that observed with filamentous actin, the liposome transport using actin bundles was more linear in nature and able to
David K Y Zhang et al.
Nature protocols, 15(3), 773-798 (2020-01-15)
Synthetic antigen-presenting cells (APCs) are used to mediate scalable ex vivo T-cell expansion for adoptive cell therapy. Recently, we developed APC-mimetic scaffolds (APC-ms), which present signals to T cells in a physiological manner to mediate rapid and controlled T-cell expansion.
Yanqi Yu et al.
ACS nano (2018-11-14)
Understanding the binding of nanoparticles to receptors on biomembranes is critical to the development and screening of therapeutic materials. A prevailing understanding is that multivalent ligand-receptor binding leads to slower and confined translational motion of nanoparticles. In contrast, we report
Michael Estl et al.
International journal of molecular sciences, 21(7) (2020-04-08)
Cytotoxic T lymphocytes (CTL) are key players of the adaptive immune system that target tumors and infected cells. A central step to that is the formation of a cell-cell contact zone between the CTL and its target called an immune
Maria Tsemperouli et al.
Langmuir : the ACS journal of surfaces and colloids, 35(26), 8748-8757 (2019-06-28)
Horizontal black lipid membranes (BLMs) enable optical microscopy to be combined with the electrophysiological measurements for studying ion channels, peptide pores, and ionophores. However, a careful literature review reveals that simultaneous fluorescence and electrical recordings in horizontal BLMs have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service