Skip to Content
Merck
All Photos(1)

Key Documents

870016P

Avanti

23:2 Diyne PC [DC(8,9)PC]

Avanti Research - A Croda Brand 870016P, powder

Synonym(s):

1,2-bis(10,12-tricosadiynoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H92NO8P
CAS Number:
Molecular Weight:
914.28
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 1 g (870016P-1g)
pkg of 1 × 200 mg (870016P-200mg)
pkg of 1 × 25 mg (870016P-25mg)
pkg of 1 × 500 mg (870016P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870016P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCC#CC#CCCCCCCCCCC)=O)COC(CCCCCCCCC#CC#CCCCCCCCCCC)=O)=O

InChI

1S/C54H92NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38-40-42-44-46-53(56)60-50-52(51-62-64(58,59)61-49-48-55(3,4)5)63-54(57)47-45-43-41-39-37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h52H,6-23,32-51H2,1-5H3/t52-/m1/s1

InChI key

IDBJTPGHAMAEMV-OIVUAWODSA-N

General description

23:2 Diyne PC [DC(8,9)PC] is a chiral diynoic phosphotidylcholine (PC).[1]

Application

23:2 Diyne PC [DC(8,9)PC] has been used in the fabrication of polydiacetylene vesicles.[2] It has also been used in the preparation of lipid-tissue plasminogen activator (lipid-tPA) suspensions.[3]

Packaging

20 mL Amber Glass Screw Cap Vial (870016P-1g)
20 mL Amber Glass Screw Cap Vial (870016P-500mg)
5 mL Amber Glass Screw Cap Vial (870016P-200mg)
5 mL Amber Glass Screw Cap Vial (870016P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Simona Colantonio et al.
    Lipids, 46(5), 469-477 (2011-02-18)
    Since its introduction as an ionization technique in mass spectrometry, matrix-assisted laser desorption ionization (MALDI) has been applied to a wide range of applications. Quantitative small molecule analysis by MALDI, however, is limited due to the presence of intense signals
    Mechanosensitivity of polydiacetylene with a phosphocholine headgroup
    Ortuso R D, et al.
    Soft Matter, 13(8), 1728-1736 (2017)
    Chiral tubule self-assembly from an achiral diynoic lipid
    Pakhomov S, et al.
    Proceedings of the National Academy of Sciences of the USA, 100(6), 3040-3042 (2003)
    Preparation of tPA-loaded microbubbles as potential theranostic agents: A novel one-step method via coaxial electrohydrodynamic atomization technique
    Yan W C, et al.
    Chemical Engineering Journal, 307(8), 168-180 (2017)
    James W Hindley et al.
    Nature communications, 9(1), 1093-1093 (2018-03-17)
    Cell-sized vesicles have tremendous potential both as miniaturised pL reaction vessels and in bottom-up synthetic biology as chassis for artificial cells. In both these areas the introduction of light-responsive modules affords increased functionality, for example, to initiate enzymatic reactions in

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service