Skip to Content
Merck
All Photos(1)

Key Documents

860837P

Avanti

CER7-2′R,6R(d9)

Avanti Research - A Croda Brand 860837P, powder

Synonym(s):

N(2′-(R)-hydroxypalmitoyl(d9)) 6R-hydroxysphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H58D9NO5
CAS Number:
Molecular Weight:
578.95
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860837P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860837P

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCC[C@@H](O)/C=C/[C@@H](O)[C@@H](NC([C@H](O)CCCCCCCCCCC([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])=O)CO

InChI key

FDCYWFADVRIZMH-LZVCNZPASA-N

General description

6-Hydroxysphingosine is one of the moieties that make up the class of ceramides, which are lipids localized to stratum corneum. It is formed due to incorporation of hydroxyl group to long chain bases.

Application

N(2′-(R)-hydroxypalmitoyl(d9)) 6R-hydroxysphingosine (CER7-2′R,6R(d9)) has been used as an internal standard for qualitative analysis of stratum corneum (SC) ceramides in humans and mice by liquid chromatography–mass spectrometry (LC-MS)/MS.

Biochem/physiol Actions

6-Hydroxysphingosine is one of the sphingoid bases in skin.

Packaging

5 mL Amber Glass Screw Cap Vial (860837P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of lipids of the stratum corneum by high performance thin layer chromatography and mass spectrometry
Jamin EL, et al.
Eur. J. Mass Spectrom., 2018, 1469066718815380-1469066718815380 (2018)
Principles and Practice of Skin Toxicology, 21-21 (2008)
Lipids and the Permeability and Antimicrobial Barriers of the Skin
Wertz Philip W
Journal of Lipids, 2018 (2018)
Momoko Kawana et al.
Journal of lipid research, 61(6), 884-895 (2020-04-09)
Ceramides are the predominant lipids in the stratum corneum (SC) and are crucial components for normal skin barrier function. Although the composition of various ceramide classes in the human SC has been reported, that in mice is still unknown, despite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service