Skip to Content
Merck
All Photos(1)

Key Documents

860626P

Avanti

C8 Dihydroceramide (d18:0/8:0)

Avanti Research - A Croda Brand 860626P, powder

Synonym(s):

N-octanoyl-D-erythro-sphinganine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H53NO3
CAS Number:
Molecular Weight:
427.70
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860626P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860626P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCC)=O)[C@]([H])(O)CCCCCCCCCCCCCCC

InChI

1S/C26H53NO3/c1-3-5-7-9-10-11-12-13-14-15-16-18-19-21-25(29)24(23-28)27-26(30)22-20-17-8-6-4-2/h24-25,28-29H,3-23H2,1-2H3,(H,27,30)/t24-,25+/m0/s1

InChI key

LGOFBZUQIUVJFS-LOSJGSFVSA-N

Application

C8 Dihydroceramide (d18:0/8:0) has been used as an internal standard to quantify the lipid extracted from human plasma by liquid chromatography mass spectrometry during high-throughput plasma lipidomics.

Biochem/physiol Actions

C8 Dihydroceramide (d18:0/8:0) or N-octanoyl-D-erythro-sphinganine, is a synthetic dihydroceramide. Dihydroceramide acts as a precursor for de-novo ceramide synthesis. C8 Dihydroceramide mimics γ-tocotrienol (γTE) in stimulating NF-κB-target protein A20 expression, increasing endoplasmic reticulum (ER) stress and reducing tumor necrosis factor (TNF)-triggered nuclear factor kappa B (NF-κB) activation.

Packaging

5 mL Amber Glass Screw Cap Vial (860626P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun Wang et al.
Journal of immunology (Baltimore, Md. : 1950), 195(1), 126-133 (2015-05-24)
NF-κB plays a central role in pathogenesis of inflammation and cancer. Many phytochemicals, including γ-tocotrienol (γTE), a natural form of vitamin E, have been shown to inhibit NF-κB activation, but the underlying mechanism has not been identified. In this study
Vitamin E gamma-tocotrienol inhibits cytokine-stimulated NF-kappaB activation by induction of anti-inflammatory A20 via stress adaptive response due to modulation of sphingolipids
Wang Y, et al.
Journal of Immunology, 195(1), 126-133 (2015)
Jennifer T Saville et al.
Journal of inherited metabolic disease, 43(3), 558-563 (2019-11-11)
Gaucher disease (GD) is an inherited metabolic disorder characterised by impaired catabolism of the glycosphingolipid, glucosylceramide. The deacetylated derivative, glucosylsphingosine (GluSph, lyso-Gb1) has materialised as a biomarker for GD. Further appraisal of the clinical utility of GluSph is required in
Low serum levels of (dihydro-) ceramides reflect liver graft dysfunction in a real-world cohort of patients post liver transplantation
Mucke V, et al.
International Journal of Molecular Sciences, 19(4), 991-991 (2018)
Victoria Therese Mücke et al.
International journal of molecular sciences, 19(4) (2018-03-29)
Patients after orthopic liver transplantation (OLT) are at risk of developing graft dysfunction. Sphingolipids (SL's) have been identified to play a pivotal role in the regulation of hepatocellular apoptosis, inflammation and immunity. We aimed to investigate the serum SL profile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service