Skip to Content
Merck
All Photos(1)

Key Documents

860492P

Avanti

Sphingosine-1-Phosphate (d18:1)

Avanti Research - A Croda Brand

Synonym(s):

D-erythro-sphingosine-1-phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H38NO5P
CAS Number:
Molecular Weight:
379.47
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 1 mg (860492P-1mg)
pkg of 1 × 25 mg (860492P-25mg)
pkg of 1 × 5 mg (860492P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@]([NH3+])([H])COP([O-])(O)=O

InChI

1S/C18H38NO5P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(20)17(19)16-24-25(21,22)23/h14-15,17-18,20H,2-13,16,19H2,1H3,(H2,21,22,23)/b15-14+/t17-,18+/m0/s1

InChI key

DUYSYHSSBDVJSM-KRWOKUGFSA-N

General description

Sphingosine-1-phosphate (S1P), a sphingolipid, is an important cellular metabolite and signaling lipid in animal cells. It has also been found in yeasts and plants.[1] Sphingosine kinases (SPKs) catalyzes the synthesis of S1P from spingosine[2] and regulates S1P levels.[3] It is the end product of ceramide catabolism and is metabolized to phosphoethanolamine and hexadecenal by S1P lyase. S1P is produced in endothelial cells.[4]

Application

Sphingosine-1-Phosphate (d18:1) has been used:
  • as an internal standard in high performance liquid chromatography (HPLC) for the quantification of plasma lipid sphingosine-1-phosphate(S1P)[5]
  • to evaluate its chemotactic response of auto reactive immature B cells[6]
  • to test its effect on autophagy and preconditioning in oxygen and glucose deprived neurons[3]

Biochem/physiol Actions

Sphingosine 1-phosphate (S1P) is a signaling molecule and an intracellular messenger, crucial for the physiological processes. It is implicated in the pathogenesis of osteoporosis, cancer, atherosclerosis and diabetes.[2] S1P regulates skin dendritic cells and keratinocytes, and its anti-inflammatory functionality is useful in treating atopic dermatitis.[4] S1P coordinates T lymphocytes circulation in lymph, plasma and tissue by interacting with G-protein coupled receptors (GPCRS).[6] S1P is a high affinity ligand for sphingosine-1-phosphate receptor (S1P1−5).[7]

Packaging

5 mL Amber Glass Screw Cap Vial (860492P-1mg)
5 mL Amber Glass Screw Cap Vial (860492P-25mg)
5 mL Amber Glass Screw Cap Vial (860492P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preconditioning stimuli induce autophagy via sphingosine kinase 2 in mouse cortical neurons
Sheng R, et al.
The Journal of Biological Chemistry, 289(30), 20845-20857 (2014)
Liao-Liao Hu et al.
Cells, 8(11) (2019-11-17)
CCN1 and CCN2 are members of the CCN family and play essential roles in the regulation of multiple female reproductive functions, including ovulation. Cyclooxygenase-2 (COX2) is a critical mediator of ovulation and can be induced by sphingosine-1-phosphate (S1P) through the
Collin Tran et al.
Journal of neurochemistry, e14917-e14917 (2019-11-20)
Sphingosine 1-phosphate (S1P) is an essential lipid metabolite that signals through a family of five G protein-coupled receptors, S1PR1-S1PR5, to regulate cell physiology. The multiple sclerosis drug Fingolimod (FTY720) is a potent S1P receptor agonist that causes peripheral lymphopenia. Recent
Effects of apolipoprotein M in uremic atherosclerosis
Bosteen MH, et al.
Atherosclerosis, 265(3), 93-101 (2017)
Suwon Jeon et al.
Journal of lipid research, 61(1), 20-32 (2019-11-07)
Sphingosine 1-phosphate (S1P) lyase is an intracellular enzyme that catalyzes the irreversible degradation of S1P and has been suggested as a therapeutic target for the treatment of psoriasis vulgaris. Because S1P induces differentiation of keratinocytes, we examined whether modulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service