Skip to Content
Merck
All Photos(2)

Documents

857228P

Avanti

C18 LPA

1-O-octadecyl-2-hydroxy-sn-glycero-3-phosphate (ammonium salt), powder

Synonym(s):

1-octadecyl-sn-glycero-3-phosphate (ammonium salt); PA(18:0e/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H48NO6P
CAS Number:
Molecular Weight:
441.58
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (LPA; may contain up to 10% of the 2-LPA isomer)

form

powder

packaging

pkg of 1 × 1 mg (857228P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857228P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(O)=O)([H])COCCCCCCCCCCCCCCCCCC.[NH4+]

General description

Lysophosphatidic acid (LPA) is a class of phospholipids, which is water-soluble. It has a glycerol backbone attached via a phosphate group, an aliphatic chain and a hydroxyl group. The aliphatic chain is linked to either sn-1 or sn-2 position and the phosphate group is connected to the sn-3 position. The hydroxyl group is linked to the remaining sn-1 or sn-2 position.

Application

C18 LPA or 1-O-octadecyl-2-hydroxy-sn-glycero-3-phosphate may be used:
  • to study its effects on Yes-associated protein (YAP) phosphorylation
  • as a standard for the quantification of lysophosphatidic acid (LPA) compounds in proton samples using reversed-phase ultra-performance liquid chromatography-tandem mass spectrometer (UPLC-MS/MS)
  • as a substrate to investigate the phosphatase activity of soluble epoxide hydrolase (sEH) towards LPA compounds

Biochem/physiol Actions

Lysophosphatidic acid (LPA) is a signaling molecule and alters various cellular responses through G protein-coupled receptors (GPCRs). It stimulates smooth muscle contraction, cytoskeleton rearrangement and chemotaxis. LPA is also involved in inducing neurotransmitter release, cell proliferation, platelet aggregation and Ca2+ mobilization. Increased levels of LPA in human plasma leads to ovarian cancer and atherosclerosis. LPA acts as a potential biomarker for ovarian cancer.

Packaging

5 mL Amber Glass Screw Cap Vial (857228P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ami Oguro et al.
Journal of lipid research, 53(3), 505-512 (2012-01-06)
Soluble epoxide hydrolase (sEH) is a bifunctional enzyme that has a C-terminus epoxide hydrolase domain and an N-terminus phosphatase domain. The endogenous substrates of epoxide hydrolase are known to be epoxyeicosatrienoic acids, but the endogenous substrates of the phosphatase activity
Lian Shan et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 864(1-2), 22-28 (2008-02-12)
Lysophosphatidic acid (LPA) is a class of lipids that play multiple biological functions. Several reports show that they are potential biomarkers for diagnosing ovarian cancer. Therefore, it is necessary to accurately quantify their levels in biological samples. Here we report
Selma Maric et al.
Applied microbiology and biotechnology, 99(1), 241-254 (2014-10-11)
Phosphatidylcholine (PC) is a major component of eukaryotic cell membranes and one of the most commonly used phospholipids for reconstitution of membrane proteins into carrier systems such as lipid vesicles, micelles and nanodiscs. Selectively deuterated versions of this lipid have
Jesica R Williams et al.
The Journal of biological chemistry, 284(25), 17304-17319 (2009-04-16)
Lysophosphatidic acid (LPA) is a ligand for LPA(1-3) of the endothelial differentiation gene family G-protein-coupled receptors, and LPA(4-8) is related to the purinergic family G-protein-coupled receptor. Because the structure-activity relationship (SAR) of GPR92/LPA(5) is limited and whether LPA is its
Han Han et al.
Molecular cell, 72(2), 328-340 (2018-10-09)
The Hippo pathway plays a crucial role in organ size control and tumor suppression, but its precise regulation is not fully understood. In this study, we discovered that phosphatidic acid (PA)-related lipid signaling is a key regulator of the Hippo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service