850700P
Avanti
10:0 PE
1,2-didecanoyl-sn-glycero-3-phosphoethanolamine, powder
Synonym(s):
PE(10:0/10:0)
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 25 mg (850700P-25mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 850700P
lipid type
phospholipids
cardiolipins
shipped in
dry ice
storage temp.
−20°C
SMILES string
[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCCCC)=O)COC(CCCCCCCCC)=O
InChI key
KKOSJVWUOHEQKA-HSZRJFAPSA-N
General description
Phosphatidylethanolamine is an aminophospholipid, which is found in eukaryotic and prokaryotic cells. It is one of the most abundant glycerophospholipids in the biological membranes. Phosphatidylethanolamine is found in nervous tissue.
Application
10:0 PE has been used for the synthesis of fluorescent phospholipid derivative NBD-didecanoylphosphatidylethanolamine (NBD-diC10PE).
Biochem/physiol Actions
Phosphatidylethanolamine forms pure lipid structures. It maintains membrane fluidity in eukaryotic cells. Phosphatidylethanolamine plays a critical role in autophagy, cell division and protein folding. It acts as a precursor for the synthesis of various protein modifications. Phosphatidylethanolamine functions as an intermediate for the synthesis of glycerophospholipid classes.
Packaging
5 mL Clear Glass Sealed Ampule (850700P-25mg)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Binding of phospholipids to beta-Lactoglobulin and their transfer to lipid bilayers
Biochimica et Biophysica Acta - Biomembranes, 1778(5), 1308-1315 (2008)
An Introduction to Biological Membranes: Composition, Structure and Function (2016)
The Membranes of Cells (2016)
Formation and function of phosphatidylserine and phosphatidylethanolamine in mammalian cells
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1831, 543-554 (2013)
Phosphatidylethanolamine and phosphatidylcholine biosynthesis by the Kennedy pathway occurs at different sites in Trypanosoma brucei
Scientific reports, 5, 16787-16787 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service