Skip to Content
Merck
All Photos(2)

Key Documents

850311C

Avanti

DOCP

Avanti Research - A Croda Brand 850311C

Synonym(s):

2-((2,3-bis(oleoyloxy)propyl)dimethylammonio)ethyl hydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H82NO8P
CAS Number:
Molecular Weight:
772.09
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850311C-25mg)
pkg of 2 × 4 mL (850311C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850311C

concentration

10 mg/mL (850311C-25mg)
25 mg/mL (850311C-200mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

DOCP is a zwitterionic /negatively charged lipid. It possess an inverse headgroup This synthetic molecule has stronger affinity than 1,2-dioleoyl-sn-glycero-3-phospho-L-serine (DOPS).
Inverse-phosphocholine lipids, a remix of a common phospholipid, contain headgroups with an inverted charge orientation relative to phosphocholine (PC) lipids. The iPC lipid headgroup has a quaternary amine adjacent to the bilayer interface and a phosphate that extends into the aqueous phase. Therefore iPC lipids afford a unique opportunity to investigate the biophysical and bioactivity-related ramifications of a charge inversion at the bilayer surface.

Application

DOCP is suitable for use in the preparation of large unilamellar vesicles (LUVs) and in the preparation of liposomes.

Packaging

5 mL Clear Glass Sealed Ampule (850311C-200mg)
5 mL Clear Glass Sealed Ampule (850311C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Partition of Positively and Negatively Charged Tryptophan Ions in Membranes with Inverted Phospholipid Heads: Simulations and Experiments
Cardenas AE, et al.
The Journal of Physical Chemistry B (2019)
A Stable Lipid/TiO2 Interface with headgroup-inversed phosphocholine and a comparison with SiO2
Wang F and Liu J
Journal of the American Chemical Society, 137(36), 11736-11742 (2015)
Charge and Coordination Directed Liposome Fusion onto SiO2 and TiO2 Nanoparticles
Wang X, et al.
Langmuir, 35(5), 1672-1681 (2018)
Inorganic cadmium affects the fluidity and size of phospholipid based liposomes
Kerek EM and Prenner EJ
Biochimica et Biophysica Acta - Biomembranes, 1858(12), 3169-3181 (2016)
Yibo Liu et al.
Langmuir : the ACS journal of surfaces and colloids, 34(32), 9337-9348 (2018-03-13)
Phospholipids are a major component of the cell membrane. In most natural phospholipids, the phosphate acts as a bridge, connecting the other portion of the polar headgroup with the hydrophobic tails. Such bridging phosphate is chemically quite inert. Synthetic lipids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service