Skip to Content
Merck
All Photos(3)

Key Documents

840051P

Avanti

Egg PC

Avanti Research - A Croda Brand

Synonym(s):

Egg PC 99%

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

description

L-α-phosphatidylcholine (Egg, Chicken)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (840051P-1g)
pkg of 2 × 100 mg (840051P-200mg)
pkg of 5 × 100 mg (840051P-500mg)
pkg of 1 × 25 mg (840051P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phosphoglycerides
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C42H82NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h20-21,40H,6-19,22-39H2,1-5H3/b21-20-/t40-/m1/s1

InChI key

WTJKGGKOPKCXLL-VYOBOKEXSA-N

General description

Phosphatidylcholines (PC) are the building blocks of eukaryotic membranes. It is also found at high levels in C. elegans lipid extracts. It is a strong bilayer-forming lipid. PC is an important component of the mucosal layer of the colon.

Application

Egg PC has been used:
  • as lipid standard to evaluate the chromatographic column characteristics
  • as a component in the surfactant comixture for theαVβ3 targeted nanoparticles
  • in liposome preparation

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance. It is used to treat fat embolism. Phosphatidylcholine lowers the levels of cholesterol and triglycerides.

Packaging

5 mL Clear Glass Sealed Ampule; Argon Filled Foil Pouch (840051P-200mg)
5 mL Clear Glass Sealed Ampule; Argon Filled Foil Pouch (840051P-25mg)
5 mL Clear Glass Sealed Ampule; Argon Filled Foil Pouch (840051P-500mg)
60 mL Amber Wide Mouth Glass Bottle with Screw Cap; Argon Filled Foil Pouch (840051P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrative Medicine - E-Book (2017)
The Membranes of Cells (2016)
Detection and quantification of angiogenesis in experimental valve disease with integrin-targeted nanoparticles and 19-fluorine MRI/MRS
Waters EA, et al.
Journal of Cardiovascular Magnetic Resonance : Official Journal of the Society for Cardiovascular Magnetic Resonance, 10(1), 43-43 (2008)
Christina M Bailey et al.
Langmuir : the ACS journal of surfaces and colloids, 33(43), 11986-11997 (2017-09-28)
Supported lipid bilayers (SLBs) have been used extensively in a variety of biotechnology applications and fundamental studies exploring lipid behavior. Despite their widespread use, various physicochemical parameters have yet to be thoroughly investigated for their impact on SLB formation. In
Qualitative and quantitative characterization of protein-phosphoinositide interactions with liposome-based methods
Busse RA, et al.
Autophagy, 9(5), 770-777 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service