Skip to Content
Merck
All Photos(1)

Key Documents

810603C

Avanti

16:0-10 Doxyl PC

Avanti Research - A Croda Brand 810603C

Synonym(s):

1-palmitoyl-2-stearoyl-(10-doxyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H90N2O10P
CAS Number:
Molecular Weight:
862.19
MDL number:
UNSPSC Code:
41141825
NACRES:
NA.12

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810603C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810603C

concentration

1 mg/mL (810603C-1mg)

lipid type

ESR probes
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

Avanti′s nitroxide spin product listing is a group of compounds designed to act as membrane probes. A variety of positions down the hydrophobic chain are labeled with the nitroxide functional groups to allow probing the membrane at various depths. These compounds have been synthesized from 1-palmitoyl-2-hydroxy-sn-glycerol-3-phosphocholine with the product being purified by column chromatography. Various n-doxyl phosphocholines have been recently used as biophysical tools to elucidate membrane trafficking with phosphatidylinositol transfer proteins and as fluorescent quenchers in lipid bilayer structural studies.
Phosphocholine is considered as a precursor molecule. It is formed during the breakdown of phosphatidylcholine metabolism.

Application

16:0-10 Doxyl PC may be used in vesicles for quenching PT-(1–46)F4W fluorescence. It may also be used in vesicles to check the fluorescence emission spectra in order to measure the penetration depth of the peptides′ tryptophan residues in lipid bilayer.

Biochem/physiol Actions

Phosphatidylcholine (PC) lowers the levels of cholesterol and triglycerides.

Packaging

5 mL Clear Glass Sealed Ampule (810603C-1mg)

Preparation Note

Product use: To prevent aggregation, prepare water-based solutions of 2 mM stock solutions of n-DOXYL PCs and store in plastic. Dilute stock solutions to 0.03- 0.1 mM solutions for EPR studies. For liposome preparations in fluorescent quenching measurements, dissolve the doxyl lipid in 150 μl absolute ethanol for a concentration of 40.3 mM , Additional supplemental information.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biology-cancer metabolic phenotype
NMR Metabolomics in Cancer Research, 15-138 (2013)
Q Wang et al.
Biochimica et biophysica acta, 1324(1), 69-75 (1997-02-21)
The N-terminal signal sequence of glucitol permease of Escherichia coli (Gut22: MIETITPGAVWFIGLFQKGGEC) and its analog (Gut22Ana: MIETITHGAEWFIGLFQKGGEC) were synthesized. The analog had a Pro residue substituted for the His at the 7th position of Gut22 and a Val residue substituted
L A Falls et al.
The Journal of biological chemistry, 276(26), 23895-23902 (2001-04-20)
The hydrophobic omega-loop within the prothrombin gamma-carboxyglutamic acid-rich (Gla) domain is important in membrane binding. The role of this region in membrane binding was investigated using a synthetic peptide, PT-(1-46)F4W, which includes the N-terminal 46 residues of human prothrombin with
Phosphatidylserine dynamics in cellular membranes.
Kay, J.G
Molecular Biology of the Cell, 23, 2198-2212 (2012)
ESR of spin-labeled bacteriophage M13 coat protein in mixed phospholipid bilayers
de Jongh HH
Biochimica et Biophysica Acta, 1024, 82-88 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service