Skip to Content
Merck

810385P

Avanti

TopFluor TMR cholesterol

Avanti Research - A Croda Brand

Synonym(s):

22-((4,4-difluoro-1,3-dimethyl-5-(4-methoxyphenyl)-4-bora-3a,4a-diaza-s-indacene-2-propionyl)amino)-23-norcholesterol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C43H56BF2N3O3
CAS Number:
Molecular Weight:
711.73
MDL number:
NACRES:
NA.25
UNSPSC Code:
12352211
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

TopFluor TMR cholesterol, Avanti Research - A Croda Brand

assay

99% (TLC)

form

powder

packaging

package of 1 × 1 mg

manufacturer/tradename

Avanti Research - A Croda Brand

fluorescence

~-545 nm~-571 nm

shipped in

dry ice

storage temp.

−20°C

General description

TopFluor (also referred to as BODIPY) is an extremely versatile fluorophore that can be structurally modified to fulfil a variety of roles. This TopFluor labeled cholesterol can be used as a tracker in a variety of biochemical and lipids research.

Packaging

5 mL Amber Glass Screw Cap Vial (810385P-1MG)

Legal Information

TopFluor is a trademark of Avanti Polar Lipids, LLC
Avanti Research is a trademark of Avanti Polar Lipids, LLC


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library