Skip to Content
Merck
All Photos(2)

Key Documents

810332P

Avanti

18:1 PE CF

Avanti Research - A Croda Brand 810332P, powder

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(carboxyfluorescein) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H94N3O14P
CAS Number:
Molecular Weight:
1136.40
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810332P-1mg)
pkg of 5 × 1 mg (810332P-5mg)
pkg of 5 × 2 mg (810332P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810332P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylethanolamine (PE) is a glycerophospholipid. 1,2-dioleyl-sn-glycero-3-phosphoethanolamine-N-carboxyfluorescein (CFPE) is a two-chain lipid. It is a fluorescent phospholipid, which carries a fluorophore in its headgroups.

Application

18:1 PE CF may be used:
  • to prepare fluorescent-labeled large unilamellar vesicles (LUV)
  • in small unilamellar lipid vesicles (SUVs) to increase fluorescence
  • to prepare fluorescent liposomes
  • to fabricate fluorescent nanomicelles

Biochem/physiol Actions

Phosphatidylethanolamine (PE) plays a vital role in the structure and function of eukaryotic membranes.

Packaging

5 mL Amber Glass Screw Cap Vial (810332P-10mg)
5 mL Amber Glass Screw Cap Vial (810332P-1mg)
5 mL Amber Glass Screw Cap Vial (810332P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaolong Huang et al.
World journal of surgical oncology, 15(1), 167-167 (2017-09-01)
Lung cancer stem cells (CSCs) are considered to be the seed of lung cancer, and CD133 is a marker of lung CSCs. Here, we developed gefitinib-loaded poly(ethylene glycol) 2000-distearoylphosphatidylethanolamine nanomicelles with CD133 aptamers (M-Gef-CD133) to eliminate CD133+ lung CSCs. M-Gef-CD133
Indrani Chakraborty et al.
Nanoscale, 9(23), 7814-7821 (2017-05-05)
The miniaturization of machines towards the micron and nanoscale requires the development of joint-like elements that enable and constrain motion. We present a facile method to create colloidal joints, that is, anisotropic colloidal particles functionalized with surface mobile DNA linkers
Danielle Hirsch-Lerner et al.
Biochimica et biophysica acta, 1714(2), 71-84 (2005-07-30)
Lipoplexes, which are complexes between cationic liposomes (L+) and nucleic acids, are commonly used as a nucleic acid delivery system in vitro and in vivo. This study aimed to better characterize cationic liposome and lipoplex electrostatics, which seems to play
Yueting Wu et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 103, 982-988 (2018-05-02)
Peripheral vascular disease (PVD) is a prevalent vascular disease that affect a large number of patients. The establishment of optimal treatments to mitigate the intimal hyperplasia (IH)-induced restenosis would help relieve the health burden of the PVD. Ribonucleotide reductase M2
Yueheng Zhang et al.
Langmuir : the ACS journal of surfaces and colloids, 35(47), 15335-15343 (2019-11-07)
Peptoids are highly biocompatible pseudopeptidic polyglycines with designable substituents on the nitrogen atoms. The therapeutic and drug-carrying potential of these materials requires a fundamental understanding of their interactions with lipid bilayers. In this work, we use amphiphilic polypeptoids with up

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service