Skip to Content
Merck
All Photos(2)

Key Documents

810176P

Avanti

18:1-12:0 NBD PA

Avanti Research - A Croda Brand 810176P, powder

Synonym(s):

1-oleoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H68N5O11P
CAS Number:
Molecular Weight:
813.96
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810176P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810176P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidic acid (PA) is an anionic cellular glycerophospholipid constituting 1-4% of membrane lipids. It possesses a phosphomonoester polar head group and two long hydrophobic acyl chains. NBD PA is a fluorescently labelled lipid with the fluorophore, 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) at the sn-2 fatty acid of PA.

Application

18:1-12:0 NBD PA or 1-oleoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphate (ammonium salt) is suitable: for the in vitro enzyme activity assay of galactolipase, for NBD-PA and NBD phosphatidyl choline uptake assays, to measure lipid transfer specificity of Ups1 mutants

Biochem/physiol Actions

Phosphatidic acid (PA) is implicated in many membrane fusion and membrane fission events, such as fission of Golgi membranes. It is a bioactive lipid and a secondary messenger.

Packaging

5 mL Amber Glass Screw Cap Vial (810176P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural determinants of lipid specificity within Ups/PRELI lipid transfer proteins
Miliara X, et al.
Nature Communications, 10(1), 1130-1130 (2019)
Lipid second messengers and Receptors
Encyclopedia of Endocrine Diseases, 77(3), 329-334 (2004)
Anh H Ngo et al.
The New phytologist, 219(1), 163-175 (2018-04-15)
Phospholipases play crucial roles in plant membrane lipid homeostasis. Nonspecific phospholipase C (NPCs) establish a unique class of phospholipases found only in plants and certain bacteria. Here, we show that two previously uncharacterized NPC isoforms, NPC2 and NPC6, are required
Xeni Miliara et al.
Nature communications, 10(1), 1130-1130 (2019-03-10)
Conserved lipid transfer proteins of the Ups/PRELI family regulate lipid accumulation in mitochondria by shuttling phospholipids in a lipid-specific manner across the intermembrane space. Here, we combine structural analysis, unbiased genetic approaches in yeast and molecular dynamics simulations to unravel
John-Paul Andersen et al.
Molecular metabolism, 42, 101055-101055 (2020-08-02)
Cholesterol plays a pivotal role in mitochondrial steroidogenesis, membrane structure, and respiration. Mitochondrial membranes are intrinsically low in cholesterol content and therefore must be replenished with cholesterol from other subcellular membranes. However, the molecular mechanisms underlying mitochondrial cholesterol transport remains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service