Skip to Content
Merck
All Photos(1)

Key Documents

800820P

Avanti

18:0 DG

1,2-dioctadecanoyl-sn-glycerol, powder

Synonym(s):

1,2-distearoyl-sn-glycerol; DG(18:0/18:0/0:0); distearin; 110882

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H76O5
CAS Number:
Molecular Weight:
625.02
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (800820P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 800820P

lipid type

neutral lipids
neutral glycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O(C(COC(=O)CCCCCCCCCCCCCCCCC)CO)C(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C39H76O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h37,40H,3-36H2,1-2H3

InChI key

UHUSDOQQWJGJQS-UHFFFAOYSA-N

General description

1,2-Distearoyl-sn-glycerol is a diacyl glycerol containing stearic acid at both the sn-1 and the sn-2 position.
In biochemical signaling, diacylglycerol (DAG) functions as a second messenger signaling lipid, and is a product of the hydrolysis of the phospholipid PIP2 (phosphatidylinositolbisphosphate) by the enzyme phospholipase C (PLC) (a membrane-bound enzyme) that, through the same reaction, produces inositol trisphosphate (IP3). Although inositol trisphosphate (IP3) diffuses into the cytosol, DAG remains within the plasma membrane due to its hydrophobic properties. IP3 stimulates the release of calcium ions from the smooth endoplasmic reticulum, whereas DAG is a physiological activator of protein kinase C (PKC). The production of DAG in the membrane facilitates translocation of PKC from the cytosol to the plasma membrane.
Diacylglycerol mimicks the effects of the tumor-promoting compounds phorbol esters.

Application

18:0 DG may be used in the preparation of liposomes for bovine serum albumin based extraction of different diacylglycerols.

Packaging

5 mL Amber Glass Screw Cap Vial (800820P-5mg)

Storage and Stability

Diacylglycerols are conveniently stored in chloroform solutions in glass vials with PTFE-lined caps at -20°C. Under these conditions acyl migration is minimal. Avoid plastic when handling chloroform solutions.

Other Notes

Delivery to cells:
Dry samples of diacylglycerol in chloroform, using a stream of nitrogen. Dissolve the residue in an appropriate volume of ethanol or DMSO, then dilute to the desired aqueous medium.
Effective concentration:
Most biological responses saturate at 20 to 250 μM sn-1,2-dioctanoylglycerol. Only sn-1,2 isomers appear to be active.
Precaution: Since short chain Diacylglycerols mimic effects of the tumor-promoting phorbol diesters in a number of biological systems, extra care should be employed in their handling. Treatment of solutions, vessels and other articles with 1N NaOH before washing or discarding will destroy diacylglycerols.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hasna Ahyayauch et al.
Biochimica et biophysica acta, 1788(3), 701-707 (2008-12-23)
Serum albumin is an abundant protein in blood plasma, that is well-known for its ability to transport hydrophobic biomolecules and drugs. Recent hypotheses propose that serum albumin plays a role in the regulation of lipid metabolism in addition to its
Calcium inhibits diacylglycerol uptake by serum albumin
Ahyayauch H, et al.
Biochimica et Biophysica Acta - Biomembranes, 1788(3), 701-707 (2009)
Specificity and Mechanism of Protein Kinase C Activation by sn-1,2-diacylglycerols.
Ganong BR, et al.
Proceedings of the National Academy of Sciences of the USA, 83, 1184-1188 (1986)
J G Ebeling et al.
Proceedings of the National Academy of Sciences of the United States of America, 82(3), 815-819 (1985-02-01)
Activation of cellular protein kinase C appears to be involved in the mechanism by which phorbol diesters induce differentiation of human myeloid leukemia cells (HL-60). Protein kinase C is thought to be physiologically activated by diacylglycerol derived from receptor-mediated phosphatidylinositol
R J Davis et al.
The Journal of biological chemistry, 260(3), 1562-1566 (1985-02-10)
The cell-permeable diacylglycerol, sn-1,2-dioctanoylglycerol (DiC8), is shown to mimic the effect of tumor promoting phorbol diesters on epidermal growth factor (EGF) binding and action in intact cells. DiC8 inhibited the binding of [3H]phorbol dibutyrate to A431 cell monolayers indicating that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service