Skip to Content
Merck
All Photos(1)

Key Documents

800100C

Avanti

18:1-2:0 DG

1-oleoyl-2-acetyl-sn-glycerol, chloroform

Synonym(s):

1--(9Z-octadecenoyl)-2-acetoyl-sn-glycerol; DG(18:1(9Z)/2:0/0:0); OAG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H42O5
CAS Number:
Molecular Weight:
398.58
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 5 mL (800100C-10mg)
pkg of 1 × 5 mL (800100C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 800100C

concentration

2 mg/mL (800100C-10mg)
5 mg/mL (800100C-25mg)

lipid type

neutral lipids
neutral glycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O([C@H](COC(=O)CCCCCCC\C=C/CCCCCCCC)CO)C(=O)C

InChI

1S/C23H42O5/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(26)27-20-22(19-24)28-21(2)25/h10-11,22,24H,3-9,12-20H2,1-2H3/b11-10-/t22-/m0/s1

InChI key

PWTCCMJTPHCGMS-YRBAHSOBSA-N

General description

18:1-2:0 DG, also called 1-oleoyl-2-acetyl-sn-glycerol (OADG), is a synthetic analog of diacyl glycerol (DAG) and is membrane permeable.

Application

18:1-2:0 DG (1-oleoyl-2-acetyl-sn-glycerol) has been used as a diacyl glycerol (DAG) analog to test its impact on DAG-sensitive channels in thalamocortical (TC) neurons. It may be used as an internal standard for retention time calibration in ultra-high performance liquid chromatography-quadrupole time-of-flight mass spectrometry (UPLC-QTOF MS) for blood sample lipid quantification. It may also be used as a protein kinase C (PKC) activator in adrenal glands.

Biochem/physiol Actions

1-oleoyl-2-acetyl-sn-glycerol (OAG) has ability to activate protein kinase C (PKC) . It also inhibits TWIK-related acid-sensitive (TASK)-like potassium channels in peripheral arterial chemoreceptors. OAG stimulates 5-lipooxygenase (5-LO) in human polymorphonuclear leukocytes (PMNL). It also inhibits mitochondrial permeability transition pore (MPTP) formation and prevents fatty acid release and necrosis in lung fibroblasts.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (800100C-10mg)
30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (800100C-25mg)

Storage and Stability

Diacylglycerols are conveniently stored in chloroform solutions in glass vials with PTFE-lined caps at -20°C. Under these conditions acyl migration is minimal. Avoid plastic when handling chloroform solutions.

Other Notes

Delivery to cells: Dry samples of diacylglycerol in chloroform, using a stream of nitrogen. Dissolve the residue in an appropriate volume of ethanol or DMSO, then dilute to the desired aqueous medium.
Effective concentration: Most biological responses saturate at 20 to 250 μM sn-1,2-dioctanoylglycerol. Only sn-1,2 isomers appear to be active.
Precaution: Since short chain Diacylglycerols mimic effects of the tumor-promoting phorbol diesters in a number of biological systems, extra care should be employed in their handling. Treatment of solutions, vessels and other articles with 1N NaOH before washing or discarding will destroy diacylglycerols.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

sn-1,2-dioctanoylglycerol: a Cell Permeable Diacylglycerol That Mimics Phorbol Diester Action on the Epidermal Growth Factor Receptor and Mitogenesis
Davis, R.J, et.al.
The Journal of Biological Chemistry, 260, 1562-1566 (1985)
Diacylglycerols and Phorbol Diesters Induce Leukemic Cell Differentiation via a Common Mechanism.
Ebeling, J.G, et.al.
Proceedings of the National Academy of Sciences of the USA, 82, 815-819 (1985)
Specificity and Mechanism of Protein Kinase C Activation by sn-1,2-diacylglycerols.
Ganong, B.R, et. al.
Proceedings of the National Academy of Sciences of the USA, 83, 1184-1188 (1986)
Exogenous sn-1,2-diacylglycerols Containing Saturated Fatty Acids Functions as Bioregulators of Protein Kinase C in Human Platelets.
Lapetina, E.G, et. al.
The Journal of Biological Chemistry, 260, 1358-1361 (1985)
Pawan Bista et al.
The Journal of physiology, 593(1), 127-144 (2015-01-06)
During the behavioural states of sleep and wakefulness thalamocortical relay neurons fire action potentials in high frequency bursts or tonic sequences, respectively. The modulation of specific K(+) channel types, termed TASK and TREK, allows these neurons to switch between the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service