Skip to Content
Merck
All Photos(1)

Key Documents

141101C

Avanti

Brain Extract Polar

Avanti Research - A Croda Brand

Synonym(s):

Brain Polar Lipid Extract (Porcine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

chloroform solution

packaging

pkg of 1 × 4 mL (141101C-100mg)
pkg of 5 × 4 mL (141101C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

25 mg/mL (141101C-100mg)
25 mg/mL (141101C-500mg)

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

Brain Extract Polar comprises phosphatidylethanolamine as major lipid component. It also contains phosphatidylserine and phosphatidylcholine. The minor components include the phosphatidic acid and phosphatidylinositol.
Brain Polar Lipid Extract is derived from the total lipid extract by precipitation with acetone followed by extraction of the acetone insoluble material with diethyl ether.

Application

Brain Extract Polar has been used as an artificial membrane solution component for permeability studies. It may be used for coating donor plate during parallel artificial membrane permeability assay (PAMPA) to study blood-brain barrier penetration. It may be used in liposome preparation for dynamin self-assembly assay.

Biochem/physiol Actions

Brain lipid, especially the polar fraction plays a key role in the inhibition of prion seeded amyloid formation. Lipid extracts from the mammalian tissues like brain, kidney and liver are used in generating model membranes for mimicking real membranes.

Packaging

5 mL Clear Glass Sealed Ampule (141101C-100mg)
5 mL Clear Glass Sealed Ampule (141101C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prediction of blood-brain barrier permeation of alpha-adrenergic and imidazoline receptor ligands using PAMPA technique and quantitative-structure permeability relationship analysis
Vucicevic J, et al.
European Journal of Pharmaceutical Sciences, 68(10), 94-105 (2015)
Chameleon-like behavior of indolylpiperidines in complex with cholinesterases targets: Potent butyrylcholinesterase inhibitors
Chierrito TPC, et al.
European Journal of Medicinal Chemistry, 145(10), 431-444 (2018)
Irene Paterniti et al.
PloS one, 6(1), e15937-e15937 (2011-02-08)
Primary traumatic mechanical injury to the spinal cord (SCI) causes the death of a number of neurons that to date can neither be recovered nor regenerated. During the last years our group has been involved in the design, synthesis and
Laura Campello et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 54(1), 142-159 (2020-02-07)
It is well established that oxidative stress and inflammation are common pathogenic features of retinal degenerative diseases. ITH12674 is a novel compound that induces the transcription factor Nrf2; in so doing, the molecule exhibits anti-inflammatory, and antioxidant properties, and affords
Endogenous brain lipids inhibit prion amyloid formation in vitro
Hoover CE, et al.
Journal of Virology, 91(9), e02162-e02116 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service