Key Documents
Z743304
Julabo® external Pt 100 bath sensors
L × diam. 600 mm × 6 mm, PTFE-coated stainless steel
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
material
PTFE-coated stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8891002
L × diam.
600 mm × 6 mm
General description
Pt 100 sensors enable you to reliably measure temperature within Julabo water baths.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)