Key Documents
Z742801
Kern EMB series precision balances
Kern EMB 2000-2, weighing capacity 2000 g, accuracy: 0.01 g, precision: ±0.05 g
Synonym(s):
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
About This Item
UNSPSC Code:
41111502
NACRES:
NB.71
packaging
pack of 1 ea
manufacturer/tradename
Kern EMB 2000-2
weighing capacity measuring range
2000 g, accuracy: 0.01 g, precision: ±0.05 g
Related Categories
General description
Entry level laboratory balance with tremendous weighing performance
  • Simple and convenient 2-key operation, making them ideal for use in schools and universities
  • Tare function facilitates formulation work
  • Particularly flat design
  • Secure and non-slip positioning with rubber feet
  • Ready for use: Batteries included. AUTO-OFF function to preserve the batteries, can be switched off
Choose from one of the most recent versions:
Certificates of Analysis (COA)