Z742336
2MAG MIXdrive 6 HT 6 point stirrer for high temperature environments, use with external control unit
Synonym(s):
induction multi-position stirrer, magnetic induction stir plate, magnetic wear-free stirrer, multi-position stirrer
About This Item
Recommended Products
material
stainless steel
description
for use with external control unit
feature
positions 6
40 W stirring power
packaging
pack of 1 ea
manufacturer/tradename
2mag 40206
parameter
≤200 °C temp. limit (in air)
≤95 °C temp. limit (in water)
W × D × H
245 mm × 375 mm × 38 mm
stirrer diam.
130 mm
stirrer volume
1-3000 mL , per stirring position
weight
8.9 kg
General description
- 2MAG MIXdrive 6 HT 6 point stirrer is ideal for routine laboratory use in chemical, biotechnical and medical applications.
- Ultra-flat, 100 % maintenance- and wear-free magnetic stirrer with inductive 2mag-Magnetic-Drive concept for powerful mixing of solutions.
- 6 stirring positions, suitable for 6 x 1,000 ml Erlenmeyer flasks, stirring point distance 130 mm.
Features and Benefits
- 100% synchronized speeds, jerk-free stirring even at low speeds.
- Fully encapsulated and hermetically sealed stainless steel housing, IP68, water-, dust-, and germproof.
- Extremely robust construction and performance, easy to clean and maximum resistance, submersible in water- or oil baths, suitable for use in CO2-incubators and ovens.
- Choice of two different temperature areas available: Basic-version up to +50 °C, HT-version up to +95 °C submerged in water baths and up to +200 °C in ovens, individual dimensions on request.
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service