Z740613
Aldrich® Essentials beaker, Berzelius
tall form, capacity 2000 mL, volume 200-2000 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
feature
subdivision 100 mL
packaging
pkg of 4 ea
manufacturer/tradename
Sigma-Aldrich
volume measuring range
200-2000 mL
capacity
2000 mL
Looking for similar products? Visit Product Comparison Guide
General description
Borosilicate glass, Berzelius, tall form beaker with reference graduations and pour spout. Standard uniform walls and flat bottom for even heat distribution.
Other Notes
Browse all glass beakers
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service