Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.76
UNSPSC Code:
41102405
AC/DC input
115 V
feature
CSA approved, for flask size 3000 mL
manufacturer/tradename
Glas-Col 100B-TM568
availability
not available in EU
output
530 watts
General description
Provides even heat for reaction kettles – ideal for resinous materials. Order heating mantle corresponding to flask manufacturer and size. Reaction flask not included.
- Not CE compliant
Other Notes
Must be used with power controller.
Legal Information
Glas-Col is a registered trademark of Glas-Col LLC
Still not finding the right product?
Explore all of our products under Glas-Col® Series TM reaction flask mantle
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.