Key Documents
Z223212
Sigma-Aldrich
Aldrich® hose clamp kits
251 pieces in assorted sizes from 1/4 to 21/4 in. o.d.
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
General description
Non-corroding, lightweight nylon clamps secure Chem-Flex and standard laboratory tubing to glassware, hose fittings, and transfer needles. Provides effective hose-to-fitting seal by squeezing clamp jaws with pliers, lateral pressure on jaws will release. Usable temperature range: −25 to 225 °F. Pressure to 150 psi.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Chem-Flex is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)