Z218650
Ace reaction flask head w/ thermometer joint
fits flask, 1,000 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Synonym(s):
flask lab, lab flask, reaction flasks
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
Joint
center joint: ST/NS24/40
side joint: ST/NS24/40
feature
fits flask 1,000 mL
manufacturer/tradename
Ace Glass 651326
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For use with reaction flasks above. Heads have three in-line joints and a #7 Ace-Thred, internally threaded side opening for thermometer (6 to 7 mm o.d.). Thread forms a compression seal with bushing and permits vertical adjustment of thermometer.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service