Key Documents
Z169064
Diamond glass cutter
nickel plated, pkg of 1 ea
Synonym(s):
Diamond cutter, Glass cutting tool, Precision glass cutter
About This Item
UNSPSC Code:
41122406
NACRES:
NB.35
Pricing and availability is not currently available.
material
nickel plated
packaging
pkg of 1 ea
Related Categories
General description
Scores/breaks single or double-strength glass. Nickel-plated brass cutter with wooden handle.
Choose from one of the most recent versions:
Certificates of Analysis (COA)