Z137308
Ace pressure filter/drying funnel
capacity 60 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
description
joint I.D. 40 mm
manufacturer/tradename
Ace Glass 719046
capacity
60 mL
disc diam.
40 mm
pore size
70-100 μm porosity
Looking for similar products? Visit Product Comparison Guide
General description
Complete with cap, funnel body, and FETFE® O-ring. Used for drying air-sensitive compounds with inert gas or as a simple pressure filter funnel. O-ring joint between cap and body gives larger opening for easier access to contents.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service